3,3',5,5'-Tetramethylbenzidine (TMB), liquid, Biotechnology Grade 1 component
Catalog # J644-100ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Abs.@650nm:<= 0.02
- pH @25°C:5.0 - 7.0
- UV/Vis Assay:PASS / Fail
Specification Test Results
About this item
:
:240.35 g/mol
:GHS08
CAS Number:54827-17-7
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748