Order Entry
Denmark
ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate Plus, liquid, Biotechnology Grade 1 component
3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate Plus, liquid, Biotechnology Grade 1 component
Catalog # K830-100ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
3,3',5,5'-Tetramethylbenzidine (TMB) membrane peroxidase substrate Plus, liquid, Biotechnology Grade 1 component
Catalog # K830-100ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Abs.@450nm (50%, 1M H2SO4):
    ≤ 0.1
  • Abs.@650nm (undiluted):
    ≤ 0.1
  • Assay (P/F):
    PASS / FAIL
  • pH @25C (undiluted):
    2.6 - 3.6

Specification Test Results

About this item

 :
 :240.35 g/mol
CAS Number:54827-17-7
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748