

2,2,4-Trimethylpentane ≥99.0% (by GC) for spectrophotometry
TCIAT0715-250ML
: TCI
:
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Absorbance (210nm):max. 0.500
- Absorbance (220nm):max. 0.200
- Absorbance (230nm):max. 0.080
:
:114.23 g/mol
:14,05193
:99°C
:–107 °C
:-9°C
:–9°C
:1262
:3,II
:GHS07
:GHS08
:540-84-1
:(CH₃)₂CHCH₂C(CH₃)₃
:MFCD00008943