Order Entry
Denmark
Orders LinkContactUsLinkComponent
Safety helmets, Concept HeightMaster
Safety helmets, Concept HeightMaster
Safety helmets, Concept HeightMaster
Certificates
Safety helmets, Concept HeightMaster
Safety helmets, Concept HeightMaster
4 Options Available Below
Product Details & Documents
Lightweight, comfortable and modern protective helmet made from ABS (acrylonitrile butadiene styrene). The Concept HeightMaster has been designed for applications where the wearer is working at height, where the suspension system and the helmet design and structure will reduce the risk of the helmet coming off as a result of an impact or a fall.
  • With extended nape area
  • Six point terylene inner cradle
  • Highly absorbent hydro-flock sweatband
  • With increased side rigidity (additional lateral deformity test)
  • Ventilation through wide ventilation slots
  • With or without ratchet, with full peak
Size: 510 to 630 mm
Weight: 325 g
Certifications: EN 397, EN 12492, EN 50365
Product Family Options
Vented, without ratchet (EU version)
Product Information
  • Colour
    Availability
    Price
  • CENUS08EKFM
    Safety helmet, vented, without ratchet, EU
    Safety helmet, vented, without ratchet, EU
    Black
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • CENUS08EWFM
    Safety helmet, vented, without ratchet, EU
    Safety helmet, vented, without ratchet, EU
    White
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • CENUS08EYFM
    Safety helmet, vented, without ratchet, EU
    Safety helmet, vented, without ratchet, EU
    Yellow
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
Vented, with ratchet (EU version)
Product Information
  • Colour
    Availability
    Price
  • CENUS08EBFMR
    Safety helmet, vented, with 'Twist 2 Fit' ratchet, EU
    Safety helmet, vented, with 'Twist 2 Fit' ratchet, EU
    Blue
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • VentPlugs, for closing ventilation holes, for Concept (vented) helmet
    VentPlugs, for closing ventilation holes, for Concept (vented) helmet
    CENUS30VP
    Log in for availability
    1 * 1 Items
Product Information
  • Catalog #
    Availability
    Price
  • Sweatband S31P, PVC, for Concept helmet
    Sweatband S31P, PVC, for Concept helmet
    CENUS31P
    Log in for availability
    1 * 1 Items - 17,00 kr