Grass snake model
Grass snake (female) model, SOMSO-Plast®, 1,1 kg, 180×180×340 mm
Catalog # SOMMZOS1033
Supplier: SOMMER MARCUS
Jump to:
Product Details & Documents
Natrix natrix.
Female grass snake with label and printed description.
Weight: 1,1 kg
: On the green base under a transparent box.
Specifications
- Weight:1,1 kg
- Description:Grass snake (female) model
- Material:SOMSO-Plast®
- W×D×H:180×180×340 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



