Order Entry
Denmark
Orders LinkContactUsLinkComponent
Viviparous lizard model

Viviparous lizard model

Viviparous lizard, SOMSO-Plast®, 1,1 kg, 180×180×340 mm
Catalog # SOMMZOS1029
Supplier:   SOMMER MARCUS
Viviparous lizard model
Catalog # SOMMZOS1029
Supplier:  SOMMER MARCUS
Viviparous lizard model
Viviparous lizard model

Viviparous lizard, SOMSO-Plast®, 1,1 kg, 180×180×340 mm

Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

Lacerta vivipara.

Includes a label and printed description.

Weight: 1,1 kg

Packaging: Under a transparent box on a green base.

Specifications
  • Weight:
    1,1 kg
  • Description:
    Viviparous lizard
  • Material:
    SOMSO-Plast®
  • W×D×H:
    180×180×340 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.