Larynx
Larynx
Catalog # SOMMGS3
Supplier: SOMMER MARCUS
Jump to:
Product Details & Documents
Cartilaginous skeleton, ligamentous apparatus, muscles, relief of mucous membrane and thyroid gland are shown. Model separates into two parts.
- Natural size
- Made in SOMSO-Plast®
- On a green base
H×W×D: 17×12×12 cm
Weight: 0,3 kg
Specifications
- Description:Larynx



