Order Entry
Denmark
ContactUsLinkComponent
EDTA (ethylenediamine tetraacetic acid) 0.1428 mol/l in aqueous solution
EDTA (ethylenediamine tetraacetic acid) 0.1428 mol/l in aqueous solution
Catalog # HACH1496001
Supplier:  Hach
CAS Number:  
undefined
EDTA (ethylenediamine tetraacetic acid) 0.1428 mol/l in aqueous solution
Catalog # HACH1496001
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :292.25 g/mol
Storage Temperature:Ambient
EINECS:200-449-4
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541