Order Entry
Denmark
ContactUsLinkComponent
 
Ethylene glycol 0,05% in aqueous solution
  HACH2183936
 :  Hach
 :  
undefined
Ethylene glycol 0,05% in aqueous solution
  HACH2183936
 :  Hach
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    15 mL
  • Seq:
    2295
  • Environmentally Preferable:

 

 

 : Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
 :
 :62.07 g/mol
 :Refrigerator
 :203-473-3
 :107-21-1
 :HOCH₂CH₂OH
 :MFCD00002885