Order Entry
Denmark
Orders LinkContactUsLinkComponent
Phenol

Phenol

Glass bottle for solids, 100 mg
Catalog # PROOMM0045.11
Toxic
Corrosive
Health
Phenol
Catalog # PROOMM0045.11
CAS Number:  108-95-2
Phenol

Glass bottle for solids, 100 mg

CAS Number:  108-95-2
Price
5.700,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Merck Index:12,07390
  • Boiling Pt:181.7 °C (1013 hPa)
  • Melting Pt:40.8 °C
  • Density:1.0722 g/cm³ (20 °C)
  • Flash Pt:81 °C
  • Storage Temperature:Ambient
  • EINECS:203-632-7
  • UN:1671
  • ADR:6.1,II
  • REACH:01-2119471329-32
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.