Order Entry
Denmark
ContactUsLinkComponent
 
Sulphuric acid 5% (v/v) in aqueous solution
  35606.K2
 :  
Sulphuric acid 5% (v/v) in aqueous solution
  35606.K2
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

 

 

 :
 :98.08 g/mol
 :13,09064
 :101 °C (1013 hPa)
 :1.066 g/cm³ (20 °C)
 :Ambient
 :231-639-5
 :3264
 :8,III
 :GHS05
 :7664-93-9
 :H₂SO₄
 :MFCD00064589