
Methyl-4-amino-3-bromobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:106896-49-5
:
Formula:C₈H₈BrNO₂
MDL Number:MFCD01861385
:230.06 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:105.0 to 109.0 deg-C
Specification Test Results
Frequently Bought Together