Organic reference standard, 1,4-Dioxane 1000 µg/ml in methanol
Catalog # CAMBULM-7840-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:123-91-1
- Description:1,4-Dioxane 1,000 µg/ml in methanol
- Formula:C₄H₈O₂
- Description:1,4-Dioxane