

Lead(II) tetrafluoroborate 50% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13814-96-5
:
Formula:B₂F₈Pb
MDL Number:MFCD00016270
:380.81 g/mol
Density:1.62
Storage Temperature:Ambient
EINECS:237-486-0
UN:2922
ADR:8,III
:GHS08
:GHS07
Related Information
Frequently Bought Together