Order Entry
Denmark
Orders LinkContactUsLinkComponent
Artificial human skeleton

Artificial human skeleton

Artificial human skeleton, SOMSO-Plast®, 10,7 kg, 1810×550×550 mm
Catalog # SOMMQS10/8
Supplier:   SOMMER MARCUS
Artificial human skeleton
Catalog # SOMMQS10/8
Supplier:  SOMMER MARCUS
Artificial human skeleton
Artificial human skeleton

Artificial human skeleton, SOMSO-Plast®, 10,7 kg, 1810×550×550 mm

Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

Natural cast of the bones of a female adult life-like representation with full anatomical detail.

  • Skull with removable vault and mandible
  • Joints movable, upper and lower limbs removable
  • Both right and left foot can be detached from the leg

Mounted upright on a stand with rollers.

Specifications
  • Weight:
    10,7 kg
  • Description:
    Artificial human skeleton
  • Material:
    SOMSO-Plast®
  • W×D×H:
    1810×550×550 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.