Section of skin
SOMMKS2
- Description:Section of skin
- Pk:1
Relief model with two hair follicles (fullyexposed and in section). Model in one piece.
- Enlarged approx. 70 times
- Made in SOMSO-Plast®
- Mounted on green board
H×W×D: 25×35×5 cm
Weight: 1,1 kg