Order Entry
Denmark
Orders LinkContactUsLinkComponent
Ear model

Ear model

Ear model, SOMSO-Plast®, 1,2 kg, 210×320×190 mm
Catalog # SOMMDS3
Supplier:   SOMMER MARCUS
Ear model
Catalog # SOMMDS3
Supplier:  SOMMER MARCUS
Ear model
Ear model

Ear model, SOMSO-Plast®, 1,2 kg, 210×320×190 mm

Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

Ear model showing tympanic membrane with malleus, incus, and labyrinth.

  • Stapes can be removed
  • Three parts enlarged three times
  • On a green base with explanations
Specifications
  • Weight:
    1,2 kg
  • Description:
    Ear model
  • Material:
    SOMSO-Plast®
  • W×D×H:
    210×320×190 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.