Order Entry
Denmark
Orders LinkContactUsLinkComponent
Capillary columns, TracePLOT™ TG-BOND S
Capillary columns, TracePLOT™ TG-BOND S
Capillary columns, TracePLOT™ TG-BOND S
Certificates
Capillary columns, TracePLOT™ TG-BOND S
3 Options Available Below
Product Details & Documents
TracePLOT™ TG-BOND S GC columns are ideal for analysis of non-polar and mid-polar compounds.
  • Mid-polarity, divinylbenzene 4-vinylpyridine solid phase
  • Particles incorporated into the walls of the tubing for essentially no particle release
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    TG-BOND S
    10,00 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.760,00 kr
    6.760,00 kr
    1 * 1 Items
       6.760,00 kr
  • Capillary
    TG-BOND S
    10,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.990,00 kr
    8.990,00 kr
    1 * 1 Items
       8.990,00 kr
  • Capillary
    TG-BOND S
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 10.100,00 kr
    10.100,00 kr
    1 * 1 Items
       10.100,00 kr