Order Entry
Denmark
Orders LinkContactUsLinkComponent
Capillary columns, TracePLOT™ TG-BOND Q+
Capillary columns, TracePLOT™ TG-BOND Q+
Capillary columns, TracePLOT™ TG-BOND Q+
Certificates
Capillary columns, TracePLOT™ TG-BOND Q+
4 Options Available Below
Product Details & Documents
Intermediate polarity columns for baseline separation of ethane, ethylene and acetylene.
  • Porous divinyl benzene homopolymer
  • Particles incorporated into the walls of the tubing for essentially no particle release
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    TG-BOND Q+
    10,00 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.760,00 kr
    6.760,00 kr
    1 * 1 Items
       6.760,00 kr
  • Capillary
    TG-BOND Q+
    10,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.830,00 kr
    8.830,00 kr
    1 * 1 Items
       8.830,00 kr
  • Capillary
    TG-BOND Q+
    20,00 µm
    0,53 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.180,00 kr
    7.180,00 kr
    1 * 1 Items
       7.180,00 kr
  • Capillary
    TG-BOND Q+
    20,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.630,00 kr
    9.630,00 kr
    1 * 1 Items
       9.630,00 kr