Shut-off valves, Upchurch Scientific®
Stop a flow stream quickly with these shut-off valves. The bodies are manufactured from either PEEK or ETFE. Both versions feature a PCTFE rotor, making them highly resistant to chemicals. The blue colorant used in some valve configurations has proven not to leach out with common HPLC solvents.
- For low and medium pressure applications
- Biocompatible, all-polymer flow path
- Pressure rated to 500 psi (34 bar)
Frequently Bought Together