Lauryl tryptose broth tubes
Katalog # HACH2162315
Supplier: Hach
CAS Number:
Specifications
- Pk:15
- Standards:-
- Description:Lauryl tryptose broth tubes
Specifications
About this item
Lauryl tryptose broth tubes, single strength, selectivity: total coliform bacteria (presumptive).
- Save time and labour costs with ready to use MPN tubes, with minimal training requirements