System for parasitological examination of stool, ParasiTrap® Eco
Supplier: Biosepar
Certificates
About this item
ParasiTrap Eco contains a filled processing and transport tube I (with Medium A Eco) and the processing tube II with integrated stainless steel filter system.
- High parasite yield and high diagnostic reliability
- Outstanding quality of microscope viewing fields
- Examinations can be interrupted at any time, including for longer periods
- No cooling, high sensitivity, environmentally friendly
- No formalin, no hazardous materials, free of ether
Accessories information: Separation medium B, staining solution medium C, Pasteur pipettes (disposable), spoons and cotton wool sticks. Additional screw caps for processing tube II.
Frequently Bought Together