Order Entry
Denmark
ContactUsLinkComponent
Mini vertical dual plate electrophoresis units
Mini vertical dual plate electrophoresis units
Supplier:  Roth Carl
Certificates
Mini vertical dual plate electrophoresis units
Supplier:  Roth Carl

Specifications

  • Catalog No:
  • ROTHY004.1
  • ROTHY003.1
  • ROTHY002.1
  • ROTHY001.1
  • Model:
  • Cooled
  • Standard
  • Cooled
  • Standard
  • Glass plate size (W×H):
  • 100×100 mm
  • 100×100 mm
  • 100×100 mm
  • 100×100 mm
  • Humidity:
  • <80 % RH
  • <80 % RH
  • <80 % RH
  • <80 % RH
  • Max. power requirement:
  • 1000 V
  • 1000 V
  • 1000 V
  • 1000 V
  • Max. temperature:
  • 65 °C
  • 65 °C
  • 65 °C
  • 65 °C
  • Spacers:
  • 1,0 mm
  • 1,0 mm
  • 1,0 mm
  • 1,0 mm
  • Sample capacity:
  • 20
  • 20
  • 20
  • 20
  • Running buffer volume:
  • 100 ml (upper tank), 300 ml (lower tank)
  • 100 ml (upper tank), 300 ml (lower tank)
  • 100 ml (upper tank), 300 ml (lower tank)
  • 100 ml (upper tank), 300 ml (lower tank)

Specifications

Combs

Product Information

Frequently Bought Together