Mop heads, DryMop™
Leverandør: MICRONOVA
Certifikater
Om denne vare
For dusting and dry mopping large surface areas.
- Low profile design
- Polyester pad head limits particle generation
- Swivel head for easy maneuvering
- Launderable/Autoclavable covers
Specifikationer
- Catalog No:
- MICRDMH-60M
- 129-0606
- 129-0607
- Environmentally Preferable:
- Reduktion af affald
Specifikationer
Frequently Bought Together