Order Entry
Denmark
ContactUsLinkComponent
VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 9 Elements in Hydrochloric Acid 10% for ICP
  84792.180
 :  
undefined
VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 9 Elements in Hydrochloric Acid 10% for ICP
  84792.180
 :  
  • Matrix:
    10% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-element quality control standard 100 mg/L {Au; Ir; Pd; Pt; Rh; Ru; Os; Sn; Te} ARISTAR® for ICP
9 Elements 100 mg/L: Au, Ir, Pd, Pt, Rh, Ru, Os, Sn, Te.
 : Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
 : Supplied with certificate of analysis.