Order Entry
Denmark
ContactUsLinkComponent
VWR® SAS Super IAQ Air Samplers
VWR® SAS Super IAQ Air Samplers
Supplier:  VWR Collection
Certificates
undefined
VWR® SAS Super IAQ Air Samplers
Supplier:  VWR Collection

Specifications

  • Catalog No:
  • 710-1048
  • 710-2149
  • Flow rate:
  • 100 L/min
  • 100 L/min
  • Sampling head:
  • Aluminium
  • Aluminium
  • Power:
  • 220 V / 50 Hz
  • 220 V / 50 Hz
  • W×D×H:
  • 105×110×290 mm
  • 105×110×290 mm
  • Weight:
  • 1,75 kg
  • 1,75 kg

Specifications

Aspirating Head

Product Information

Frequently Bought Together