Order Entry
Denmark
ContactUsLinkComponent
Air sampler, MAS-100 Eco®
Air sampler, MAS-100 Eco®
Catalog # 1.09227.0001
Supplier:  Merck
undefined
Air sampler, MAS-100 Eco®
Catalog # 1.09227.0001
Supplier:  Merck

Specifications

  • Model:
    MAS-100 Eco®
  • Flow rate:
    100 L/min
  • Description:
    Air sampler
  • Display:
    Alphanumeric LCD, with 16 digits, real time and date
  • Material:
    Silver
  • Sampling head:
    Perforated aluminium lid with 400 holes
  • Pk:
    1

Specifications

About this item

The MAS-100 Eco® is a robust and reliable air sampler designed for highest precision.

  • Uses 90 mm Petri dishes
  • Programmable delay before sampling starts and pre-selected volumes, individually adjustable
  • Small and handy, fits perfectly into a HACCP concept
  • Six pre-selected volumes
  • Operation time: 3 hours
  • Autoclavable sampling head to avoid any contamination

Suitable for air sampling in food and beverage industries.

Certifications: ISO 14698 - 1/2, EN 60079.
Delivery: Supplied in a small grey carrying case.