


(Chloromethylene)dimethylammonium chloride ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3724-43-4
:
Formula:C₃H₇Cl₂N
MDL Number:MFCD00011868
:128.00 g/mol
Storage Temperature:Refrigerator
EINECS:425-970-6
UN:2923
ADR:8,III
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Argentmetric Titration):min. 95.0 %
Specification Test Results
Frequently Bought Together