Order Entry
Denmark
ContactUsLinkComponent
Multiroll tape dispenser
Multiroll tape dispenser
  DELT191220
 :  DELTALAB
Multiroll tape dispenser
  DELT191220
 :  DELTALAB

 

  • For Use With:
    Tapes with 13 mm width
  • Description:
    Multiroll tape dispenser, for 12× 13 mm rolls
  • Pk:
    1

 

 

This 12 roll tape metal dispenser accepts 13 m rolls.