Bottles, narrow neck, borosilicate glass, clear, with screw cap
Supplier: DELTALAB
Certificates
About this item
Bottles, made of borosilicate glass 3,3. Ideal for chemical reagents and culture media.
- Autoclavable up to 140 °C
- Graduations printed in white colour
- Screw caps made of blue polypropylene
- Ribbed screw caps for a better handling
- Drip ring on bottle neck to avoid dripping
Frequently Bought Together