Order Entry
Denmark
Orders LinkContactUsLinkComponent
Capillary columns, TRACE™ TR-V1
Capillary columns, TRACE™ TR-V1
Capillary columns, TRACE™ TR-V1
Certificates
Capillary columns, TRACE™ TR-V1
Capillary columns, TRACE™ TR-V1
6 Options Available Below
Product Details & Documents
Thick film columns for the analysis of volatile analytes.
  • Mid-polarity phase, 6% cyanopropylphenyl polysiloxane
  • Low bleed suitable for MS detection
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    TR-V1
    1,00 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.840,00 kr
    6.840,00 kr
    1 * 1 Items
       6.840,00 kr
  • Capillary
    TR-V1
    1,40 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.270,00 kr
    7.270,00 kr
    1 * 1 Items
       7.270,00 kr
  • Capillary
    TR-V1
    1,40 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 12.180,00 kr
    12.180,00 kr
    1 * 1 Items
       12.180,00 kr
  • Capillary
    TR-V1
    1,80 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.640,00 kr
    7.640,00 kr
    1 * 1 Items
       7.640,00 kr
  • Capillary
    TR-V1
    1,80 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 12.770,00 kr
    12.770,00 kr
    1 * 1 Items
       12.770,00 kr
  • Capillary
    TR-V1
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.280,00 kr
    8.280,00 kr
    1 * 1 Items
       8.280,00 kr