Order Entry
Denmark
ContactUsLinkComponent
Bottle top tilt measures, with flask
Bottle top tilt measures, with flask
Catalog # KAVA632446003005
Supplier:  Kavalier
undefined
Bottle top tilt measures, with flask
Catalog # KAVA632446003005
Supplier:  Kavalier

Specifications

  • Capacity:
    5 ml
  • Description:
    Automatic pipette, Kipp, with bottle (250 ml), NS 29/32
  • Pk:
    10

Specifications

About this item

Borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

This dispenser head with standard ground jont NS 29/32 is completed by a 250 ml flask for repetitive dispensing of the same volume.