n-Decane, anhydrous 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:124-18-5
:
Formula:H₃C(CH₂)₈CH₃
MDL Number:MFCD00008954
:142.28 g/mol
Boiling Pt:174 °C (760 mmHg)
Melting Pt:–30 °C
Density:0.735 g/cm³
Flash Pt:46 °C
Storage Temperature:Ambient
EINECS:204-686-4
UN:2247
ADR:3,III
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥98.5 %
- Residue after evaporation:≤0.0005 %
Specification Test Results
Related Information
Frequently Bought Together