D-(+)-Glucose-[1-¹³C] (98-99% ¹³C )
1 g
Catalog # CAMBCLM-420-1
Supplier: CAMBRIDGE ISOTOPE LABS
D-(+)-Glucose-[1-¹³C] (98-99% ¹³C )
Catalog # CAMBCLM-420-1
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number: 40762-22-9
Jump to:
Product Details & Documents
- CAS Number:40762-22-9
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00037545
Specifications
Specifications
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



