
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) ≥98.0% (by HPLC) coupling agent for peptides synthesis
: TCI
:128625-52-5
:
:C₁₈H₂₈F₆N₆OP₂
:MFCD00077411
:520.4 g/mol
:GHS07
:Refrigerator
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
Frequently Bought Together