Order Entry
Denmark
Orders LinkContactUsLinkComponent
Woulff bottles
Woulff bottles
Woulff bottles
Woulff bottles
Woulff bottles
Made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion gives relatively high resistance to temperature changes
With two side necks GL 18, center neck GL 25, complete with screw caps.
Product Family Options
  • Capacity
    Ext.Ø
    Height
  • 500 ml
    85 mm
    155 mm
    1 * 1 Items
    1 * 1 Items - 1.400,00 kr
    1.400,00 kr
    1 * 1 Items
       1.400,00 kr
  • 1000 ml
    110 mm
    185 mm
    1 * 1 Items
    1 * 1 Items - 1.460,00 kr
    1.460,00 kr
    1 * 1 Items
       1.460,00 kr