Safety storage cabinets, type 90, Q-PEGASUS-90
Supplier: asecos
Certificates
About this item
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe, durable, robust and convenient with transport base and bicoloured cabinet design. The PEGASUS line is nearly identical to the CLASSIC line, but with integrated AGT (one handed operation for double door cabinets) and integrated TSA (door open arrest system and automatic door closing).
- Robust construction and longevity - triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch and impact resistant surface, easy to clean
- Convenient handling - one handed operation for double door cabinets (AGT)
- User friendly and safe - door open arrest system (standard), offers convenient and safe removal or storage of containers
- Always closed - serial automatic door closing (TSA) releases arrested doors after ~60 seconds. Acoustic and visual signal before closing
- No unauthorised use - doors lockable with cylinder locking (integration in a locking system possible) and locking state indicator (red/green)
- Easy transport - integrated transport base with removable, optional base cover
- Easy alignment - adjusting aids to compensate for uneven floor
- Quickly put into operation - integrated air ducts ready for connection (NW 75 mm) to a technical exhaust system
The new two-colour look with a choice of seven door colours (on request), at no extra cost, offers maximum individuality and a modern appearance.
Furnace tested (type test) in accordance with EN 14470-1
Fire resistance 90 minutes
GS approval, CE conformity
EN 14727 conformity (laboratory furniture)
Certifications: Furnace tested (type test) in accordance with EN 14470-1; fire resistance upto 90 minutes; GS approval; CE conformity; EN 14727 conformity (laboratory furniture).
Ordering information: Supplied without interior equipment packages. Interior equipment packages need to be ordered separately. Please check the website or contact Avantor for further information.
Delivery: Delivery prices available upon request.
Frequently Bought Together