Blood collection needles
Supplier: SARSTEDT
Certificates
About this item
The needle protection on the safety-needles and Safety-Multifly® needles enables the safe termination of blood collection by locking the needle in the needle protector.
Frequently Bought Together