Order Entry
Denmark
Orders LinkContactUsLinkComponent
Capillary columns, TRACE™ TR-5
Capillary columns, TRACE™ TR-5
Capillary columns, TRACE™ TR-5
Certificates
Capillary columns, TRACE™ TR-5
Capillary columns, TRACE™ TR-5
13 Options Available Below
Product Details & Documents
Excellent starting columns for method development, capable of performing most required separations.
  • Non-polar phase, 5% phenyl methyl polysiloxane
  • High operating temperature and extremely low bleed
  • Widely used in a variety of applications
Documents
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    TR-5
    0,50 µm
    0,32 mm
    100 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 13.990,00 kr
    13.990,00 kr
    1 * 1 Items
       13.990,00 kr
  • Capillary
    TR-5
    0,25 µm
    0,25 mm
    7 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.400,00 kr
    1.400,00 kr
    1 * 1 Items
       1.400,00 kr
  • Capillary
    TR-5
    0,25 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.940,00 kr
    3.940,00 kr
    1 * 1 Items
       3.940,00 kr
  • Capillary
    TR-5
    0,25 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.660,00 kr
    6.660,00 kr
    1 * 1 Items
       6.660,00 kr
  • Capillary
    TR-5
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.100,00 kr
    7.100,00 kr
    1 * 1 Items
       7.100,00 kr
  • Capillary
    TR-5
    0,25 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 11.720,00 kr
    11.720,00 kr
    1 * 1 Items
       11.720,00 kr
  • Capillary
    TR-5
    0,25 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 12.530,00 kr
    12.530,00 kr
    1 * 1 Items
       12.530,00 kr
  • Capillary
    TR-5
    0,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.000,00 kr
    8.000,00 kr
    1 * 1 Items
       8.000,00 kr
  • Capillary
    TR-5
    1,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.310,00 kr
    7.310,00 kr
    1 * 1 Items
       7.310,00 kr
  • Capillary
    TR-5
    1,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.000,00 kr
    8.000,00 kr
    1 * 1 Items
       8.000,00 kr
  • Capillary
    TR-5
    1,50 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.790,00 kr
    7.790,00 kr
    1 * 1 Items
       7.790,00 kr
  • Capillary
    TR-5
    5,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.770,00 kr
    7.770,00 kr
    1 * 1 Items
       7.770,00 kr
  • Capillary
    TR-5
    0,50 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.100,00 kr
    7.100,00 kr
    1 * 1 Items
       7.100,00 kr