Order Entry
Denmark
ContactUsLinkComponent
Spray bottles
Spray bottles
Catalog # SEMA3331
Supplier:  SEMADENI
undefined
Spray bottles
Catalog # SEMA3331
Supplier:  SEMADENI
Specifications
  • Capacity:
    550 ml
  • Colour:
    Transparent
  • Height:
    215 mm
  • Pk:
    1
Specifications
About this item
Made of HDPE.
  • Good chemical resistance
  • Good temperature resistance
  • Robust, convenient, stable
By turning the nozzle, delivery can be varied from a fine spray to a solid stream.