Order Entry
Denmark
ContactUsLinkComponent
3-Hydroxy-4-nitrobenzaldehyde ≥97.0% (by GC, titration analysis)
Irritant
3-Hydroxy-4-nitrobenzaldehyde ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
3-Hydroxy-4-nitrobenzaldehyde ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    129.0 to 133.0 deg-C

Specification Test Results

Frequently Bought Together