Order Entry
Denmark
Orders LinkContactUsLinkComponent
Paracetamol

Paracetamol

Glass bottle for solids, 100 mg
Catalog # PROOBP371
Paracetamol
Catalog # PROOBP371
CAS Number:  103-90-2
Paracetamol

Glass bottle for solids, 100 mg

CAS Number:  103-90-2
Price
3.760,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
  • CAS Number:103-90-2
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00002328
  • Molecular Weight:151.16 g/mol
  • Merck Index:13,00048
  • Boiling Pt:388 °C (1013 hPa)
  • Melting Pt:168 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-157-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications