D-(-)-Diethyl tartrate ≥97.0% (by GC)
: TCI
:13811-71-7
:
:C₈H₁₄O₆
:MFCD00064451
:14,03855
:130°C
:17 °C
:1,2 g/cm³ (20 °C)
:93°C
:206.2 g/mol
:237-458-8
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
- Specific rotation [a]20/D:-7.5 to -8.1 deg (neat)
Frequently Bought Together