Potassium sulphate ≥99.0%, fine crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7778-80-5
:
Formula:K₂SO₄
MDL Number:MFCD00011388
:174.26 g/mol
Merck Index:13,07759
Boiling Pt:1698 °C (1013 hPa)
Melting Pt:1069 °C
Density:2.66 g/cm³
Storage Temperature:Ambient
EINECS:231-915-5
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:≤0.01%
- Assay (K₂SO₄):≥ 99.0 %
- Insoluble Matter:≤ 0.01 %
- pH of 5% Solution at 25°C:5.5 - 8.5
- Chloride (Cl):≤ 0.001 %
- Calcium (Ca):≤ 0.01 %
- Magnesium (Mg):≤ 0.005 %
- Sodium (Na):≤ 0.02 %
- Nitrogen Compounds (as N):≤ 5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤ 5 ppm
- Trace Impurities - Iron (Fe):≤ 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKR3278
SDS
spec_BAKR3278-05 spec_BAKR3278-07
Product specification
Frequently Bought Together