Order Entry
Denmark
ContactUsLinkComponent
Bis-trispropane ≥99.0% (by titrimetric analysis) for buffer material
Bis-trispropane ≥99.0% (by titrimetric analysis) for buffer material
  TCIAB1057-25G
 :  TCI
 :  
undefined
Bis-trispropane ≥99.0% (by titrimetric analysis) for buffer material
  TCIAB1057-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    165.0 to 167.0 deg-C
  • Solubility in Water:
    almost transparency

 

 

 :
 :282.34 g/mol
 :264-899-3
 :64431-96-5
 :C₁₁H₂₆N₂O₆
 :MFCD00004689