Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Supplier: RESEARCH ORGANICS
Certificates
About this item
Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
CAS Number:77-86-1
:
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
:121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
EINECS:201-064-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together