
6-(Trifluoromethyl)nicotinic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:231291-22-8
:
Formula:C₇H₄F₃NO₂
MDL Number:MFCD00792430
:191.11 g/mol
Melting Pt:191…192 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together