Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Buffers, MOPS, 0,5 M solutions

Buffers, MOPS, 0,5 M solutions
 
Buffers, MOPS, 0,5 M solutions
Buffers, MOPS, 0,5 M solutions

 

 

 

 

  • Catalog No:
  • J62839.AK
  • J62839.AE
  • J60328.AE
  • J60328.AK
  • Odour/scent:
  • Odourless
  • Odourless
  • Odourless
  • Odourless
  • Physical form:
  • Liquid
  • Liquid
  • Liquid
  • Liquid
  • CAS No.:
  • 1132-61-2
  • 1132-61-2
  • 1132-61-2
  • 1132-61-2
  • MDL No.:
  • MFCD00006183
  • MFCD00006183
  • MFCD00006183
  • MFCD00006183
  • Storage temperature:
  • 2°C to 8°C
  • 2°C to 8°C

 

Product Family Options

 

  • Description
    Concentration
    pH value
    Pack type
    Pk Info
    Pk
     
     
  • MOPS buffer solution
    0,5 mol/L
    7,5
    Glass bottle
    23
    250 mL
       
       
    1 * 250 mL
    1 * 250 mL - 1.320,00 kr
    1.320,00 kr
    1 * 250 mL
       1.320,00 kr

     
     
    Specifications
    Cat. No.
    J62839.AK
    Odour/scent
    Odourless
    Physical form
    Liquid
    CAS No.
    1132-61-2
    MDL No.
    MFCD00006183
    Storage temperature
    2°C to 8°C
  • MOPS buffer solution
    0,5 mol/L
    7,5
    Glass bottle
    22
    100 mL
       
       
    1 * 100 mL
    1 * 100 mL - 888,00 kr
    888,00 kr
    1 * 100 mL
       888,00 kr

     
     
    Specifications
    Cat. No.
    J62839.AE
    Odour/scent
    Odourless
    Physical form
    Liquid
    CAS No.
    1132-61-2
    MDL No.
    MFCD00006183
    Storage temperature
    2°C to 8°C
  • MOPS buffer solution
    0,5 mol/L
    7,0
    Glass bottle
    18
    100 mL
       
       
    1 * 100 mL
    1 * 100 mL - 962,00 kr
    962,00 kr
    1 * 100 mL
       962,00 kr

     
     
    Specifications
    Cat. No.
    J60328.AE
    Odour/scent
    Odourless
    Physical form
    Liquid
    CAS No.
    1132-61-2
    MDL No.
    MFCD00006183
    Storage temperature
  • MOPS buffer solution
    0,5 mol/L
    7,0
    Glass bottle
    19
    250 mL
       
       
    1 * 250 mL
    1 * 250 mL - 1.310,00 kr
    1.310,00 kr
    1 * 250 mL
       1.310,00 kr

     
     
    Specifications
    Cat. No.
    J60328.AK
    Odour/scent
    Odourless
    Physical form
    Liquid
    CAS No.
    1132-61-2
    MDL No.
    MFCD00006183
    Storage temperature
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.