
Tris(pentafluorophenyl)borane ≥98.0% (by NMR)
Supplier: TCI
Certificates
About this item
CAS Number:1109-15-5
:
Formula:C₁₈BF₁₅
MDL Number:MFCD00269813
Merck Index:14,09755
Melting Pt:128°C
EINECS:213-504-2
ADR:6.1,III
:511.98 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (NMR):min. 98.0 atom%
- Water:max. 1.0 %
Specification Test Results
Related Information
Frequently Bought Together