
4-(4-Methylphenylthio)benzophenone ≥98.0% (by GC)
Catalog # TCIAB1267-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:83.0 to 86.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:304.4054 g/mol
EINECS:281-064-9
:GHS07
CAS Number:83846-85-9
Formula:C₂₀H₁₆OS
MDL Number:MFCD00055651